AMOXICILLIN IMPURITY D 1 * 50 mg
Catalog # PHR2191-50MG
Supplier: Merck
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Titanium trichloride sulphuric acid reagent Reag. Ph. Eur. 1091202
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Acetate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Supplier: HAMILTON BONADUZ
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Buffer solutions in sachets
Supplier: Mettler - Toledo
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
Bromophenol blue solution R1 Reag. Ph. Eur. 1012802
Supplier: VWR Chemicals
AMOXICILLIN RELATED COMPOUND E 1 * 50 mg
Supplier: Merck