Order Entry
France
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ei) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession QHD43416.1). Mutations T19R, V70F, FR157-158Del, A222V, W258L, K417N, L452R, T478K, D614G, P681 1 * 25 µG
Catalog # ACRBSPN-C82EI-25UG
Fournisseur:  ACROBIOSYSTEMS
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ei) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession QHD43416.1). Mutations T19R, V70F, FR157-158Del, A222V, W258L, K417N, L452R, T478K, D614G, P681 1 * 25 µG
Catalog # ACRBSPN-C82EI-25UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together