[EN]MOBILE T-F N96 1 * 1 Pce
Catalog # MSPP21BSPSSTF0
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Specialty pH electrodes, InLab®
Supplier: Mettler - Toledo
Accessories for PIPETBOY acu / pro
Supplier: Integra
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG