Order Entry
France
Orders LinkContactUsLinkComponent
 

(S)-(-)-2-Methoxypropionic acid 97%

Bottle, 1 g
Catalog # APOSOR923407-1G
Supplier:   Apollo Scientific
(S)-(-)-2-Methoxypropionic acid 97%
Catalog # APOSOR923407-1G
Supplier:  Apollo Scientific
CAS Number:  23953-00-6
(S)-(-)-2-Methoxypropionic acid 97%

Bottle, 1 g

CAS Number:  23953-00-6
Price
33,30 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (S)-2-Methoxypropanoic acid
  • CAS Number:23953-00-6
  • Formula:C₄H₈O₃
  • MDL Number:MFCD01632589
  • Molecular Weight:104.105 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.