Order Entry
France
Orders LinkContactUsLinkComponent
Propionic acid ≥99.7%, GPR RECTAPUR®

Propionic acid ≥99.7%, GPR RECTAPUR®

Glass bottle, 2,5 L
Catalog # 87062.320
Corrosive
Flammable
Irritant
Supplier:   VWR Chemicals
Propionic acid ≥99.7%, GPR RECTAPUR®
Catalog # 87062.320
Supplier:  VWR Chemicals
CAS Number:  79-09-4
Propionic acid ≥99.7%, GPR RECTAPUR®
Propionic acid ≥99.7%, GPR RECTAPUR®

Glass bottle, 2,5 L

CAS Number:  79-09-4
Price
86,10 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Boiling Pt:140…142 °C
  • Flash Pt:54 °C
  • Storage Temperature:Ambient
  • UN:3463
  • ADR:8,II
Specifications

Specification Test Results

  • Assay (GC) :
    min 99.0%
  • Identity (IR) :
    passes test
  • Density (d 20°C/4°C):
    0,993 – 0,994
  • Water (KF):
    max 0.30%
  • Appearance:
    clear liquid

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.