Order Entry
France
ContactUsLinkComponent
Human LIGHT, His Tag active trimer MALS verified LIT-H5242 is expressed from human 293 cells HEK293. It contains AA Asp 74 - Val 240 Accession O43557-1 trimer Design. 1 * 1 mg
Catalog # ACRBLIT-H5242-1MG
Supplier:  ACROBIOSYSTEMS
Human LIGHT, His Tag active trimer MALS verified LIT-H5242 is expressed from human 293 cells HEK293. It contains AA Asp 74 - Val 240 Accession O43557-1 trimer Design. 1 * 1 mg
Catalog # ACRBLIT-H5242-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together