Order Entry
France
ContactUsLinkComponent
Human FAP, His Tag MALS verified FAP-H5244 is expressed from human 293 cells HEK293. It contains AA Leu 26 - Asp 760 Accession Q128 84-1. 1 * 500 µG
Catalog # ACRBFAP-H5244500UG
Supplier:  ACROBIOSYSTEMS
Human FAP, His Tag MALS verified FAP-H5244 is expressed from human 293 cells HEK293. It contains AA Leu 26 - Asp 760 Accession Q128 84-1. 1 * 500 µG
Catalog # ACRBFAP-H5244500UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together