Order Entry
France
ContactUsLinkComponent
Human MICA, Fc Tag MALS verified MIA-H5253 is expressed from human 293 cells HEK293. It contains AA Glu 24- Gln 308 Accession AAH16 929.1. 1 * 100 µG
Catalog # ACRBMIA-H5253100UG
Supplier:  ACROBIOSYSTEMS
Human MICA, Fc Tag MALS verified MIA-H5253 is expressed from human 293 cells HEK293. It contains AA Glu 24- Gln 308 Accession AAH16 929.1. 1 * 100 µG
Catalog # ACRBMIA-H5253100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together