Order Entry
France
ContactUsLinkComponent
Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Chimeric mAb, Human IgG1 AM128 SPD-M128 is a chimeric monoclonal antibody combinin g the constant domains of the human IgG1 molecule with mouse variable regions. Pseudovirus assay shows that t 1 * 1 mg
Catalog # ACRBSPD-M128-1MG
Supplier:  ACROBIOSYSTEMS
Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Chimeric mAb, Human IgG1 AM128 SPD-M128 is a chimeric monoclonal antibody combinin g the constant domains of the human IgG1 molecule with mouse variable regions. Pseudovirus assay shows that t 1 * 1 mg
Catalog # ACRBSPD-M128-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together