Order Entry
France
ContactUsLinkComponent
Human M-CSF R, Mouse IgG2a Fc Tag MALS verified CSR-H5255 is expressed from human 293 cells HEK293. It contains AA Ile 20 - Glu 512 Accession P07333-1. 1 * 1 mg
Catalog # ACRBCSR-H5255-1MG
Supplier:  ACROBIOSYSTEMS
Human M-CSF R, Mouse IgG2a Fc Tag MALS verified CSR-H5255 is expressed from human 293 cells HEK293. It contains AA Ile 20 - Glu 512 Accession P07333-1. 1 * 1 mg
Catalog # ACRBCSR-H5255-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together