Model X range disposable masks, Respair®
Respair® Model X range disposable mask, valved, FFP3
Catalog # JSPAFFP3V-10
Supplier: JSP
Jump to:
Product Details & Documents
A range of vertical fold-flat disposable masks with unique bi-directional filter material, allowing freedom of movement and easy communication.
- Lightweight and individually wrapped
- Ergonomically shaped for comfort and ease of use
- Adjustable head straps provide an individual fit
- Bi-directional filter protects the wearer and the wearer's environment
- Valved and unvalved versions available
An easy fit minimises leakage to ensure a high level of protection and comfort, plus complete compatibility with all other forms of protective equipment for an especially clear field of vision. A range of protection factors are available in both valved and non-valved versions. FFP2 rated masks filter 94% of airborne particulates and FFP3 rated masks filter 99% of airborne particulates.
: EN149.
Specifications
- Description:Respair® Model X range disposable mask, valved
- Seq:04
- Class:FFP3