Order Entry
France
ContactUsLinkComponent
Heating Circulator Unistat T305w HT Temperature range:      (15) 65 ... 300 °C Heating capacity: 2,5 - 3 kW Pressure pump max.:     45 l/min / 0.9 bar Pump connection:  M24x1,5 male  Power supply:     200V-240V 1~/2~ 50/60Hz 1 * 1 items
Catalog # HUBE1003.0039.01
Supplier:  HUBER
Heating Circulator Unistat T305w HT Temperature range:      (15) 65 ... 300 °C Heating capacity: 2,5 - 3 kW Pressure pump max.:     45 l/min / 0.9 bar Pump connection:  M24x1,5 male  Power supply:     200V-240V 1~/2~ 50/60Hz 1 * 1 items
Catalog # HUBE1003.0039.01
Supplier:  HUBER
Frequently Bought Together