AVP ELISA Kit (Rat)is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for AVP has been pre-coated onto a 96-well plate (12 x 8 Well Strips) and blocked. 1 * 1 KIT
Catalog # AVIVOKCD06267
Supplier: Aviva Systems Biology
Frequently Bought Together