Order Entry
France
ContactUsLinkComponent
MicroGenesis Single Use Microfiber mop cover with pockets, 12.7 x 39.3cm, use with Contec Mop Head Saturation System (2787 or 2940P3SD or 2900) and mop frame 2901 24packs x 5pcs irradiated 1 * 120 items
Catalog # CONTMGMP0005
Supplier:  CONTEC
MicroGenesis Single Use Microfiber mop cover with pockets, 12.7 x 39.3cm, use with Contec Mop Head Saturation System (2787 or 2940P3SD or 2900) and mop frame 2901 24packs x 5pcs irradiated 1 * 120 items
Catalog # CONTMGMP0005
Supplier:  CONTEC
Frequently Bought Together