POTASSIUM PYROANTIMONATE TS. DUREE DE CO 1 * 100 mL
Catalog # ACSDUSP299
Supplier: CPACHEM
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR® ARISTAR®, Sodium 1000 mg/L in Water Standard for Ion Chromatography
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Propionic anhydride ≥97%
Supplier: Thermo Fisher Scientific