Order Entry
France
ContactUsLinkComponent
Biotinylated 2019-nCoV (COVID-19) S1 protein, His,Avitag™ (MALS verified) Biotinylated 2019-nCoV (COVID-19) S1 protein, His,Avitag ( MALS verified) (S1N-C82E8) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession Q s 1 * 200 µG
Catalog # ACRBS1N-C82E8200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated 2019-nCoV (COVID-19) S1 protein, His,Avitag™ (MALS verified) Biotinylated 2019-nCoV (COVID-19) S1 protein, His,Avitag ( MALS verified) (S1N-C82E8) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession Q s 1 * 200 µG
Catalog # ACRBS1N-C82E8200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together