Order Entry
France
ContactUsLinkComponent
Biotinylated 2019-nCoV (COVID-19) S protein RBD, His,Avitag™ (MALS verified) Biotinylated 2019-nCoV (COVID-19) S protein RBD, His,Av itag (MALS verified) (SPD-C82E9) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Access 1 * 200 µG
Catalog # ACRBSPD-C82E9200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated 2019-nCoV (COVID-19) S protein RBD, His,Avitag™ (MALS verified) Biotinylated 2019-nCoV (COVID-19) S protein RBD, His,Av itag (MALS verified) (SPD-C82E9) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Access 1 * 200 µG
Catalog # ACRBSPD-C82E9200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together