Order Entry
France
Orders LinkContactUsLinkComponent
Eletriptan Related Compounds, Reference Standard

Eletriptan Related Compounds, Reference Standard

Eletriptan Hydrobromide, USP reference standard, Vial, 200 mg
Catalog # 1234453.
Supplier:   USP
Eletriptan Related Compounds, Reference Standard
Catalog # 1234453.
Supplier:  USP
CAS Number:  
Eletriptan Related Compounds, Reference Standard

Eletriptan Hydrobromide, USP reference standard, Vial, 200 mg

CAS Number:  
Price
675,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Eletriptan Hydrobromide
  • Cat. No.:
    1234453.
  • Description:
    Eletriptan Hydrobromide
  • Synonym(s):
    3-[[(R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]indole monohydrobromide monohydrate
  • CAS No.:
    273211-28-2

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.