ActiveMax® Human VEGF165, Tag Free (MALS verified) (VE5-H4210) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 191 (Accession P15692-4). 1 * 20 µG
ACRBVE5-H4210-20UG
ActiveMax® Human VEGF165, Tag Free (MALS verified) (VE5-H4210) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 191 (Accession P15692-4). 1 * 20 µG
ACRBVE5-H4210-20UG
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Stoppers for standard ground joints, octogonal, HDPE
: GLASWARENFABRIK KARL HECHT
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
ClipTip™ Pipette Tips
: Thermo Fisher Scientific
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF
VWR® Reagent Reservoirs
: VWR Collection