Order Entry
France
ContactUsLinkComponent
MABSol® Biotinylated Human CD3ECD3D Heterodimer Protein, Fc,His TagFc,Flag Tag, primary amine labeling (CDD-H82W0) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (CD3E) Phe 22 - Ala 105 (CD3D) (Accession NP000724.1 (CD3E) NP00 1 * 200 µG
Catalog # ACRBCDD-H82W0200UG
Supplier:  ACROBIOSYSTEMS
MABSol® Biotinylated Human CD3ECD3D Heterodimer Protein, Fc,His TagFc,Flag Tag, primary amine labeling (CDD-H82W0) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (CD3E) Phe 22 - Ala 105 (CD3D) (Accession NP000724.1 (CD3E) NP00 1 * 200 µG
Catalog # ACRBCDD-H82W0200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together