MABSol® Biotinylated Human TNF-alpha, His,Avitag (active trimer) (MALS verified) (TNA-H82E3) is expressed from human HEK293 cells. I t contains AA Val 77 - Leu 233 (Accession NP000585.2). 1 * 200 µG
Catalog # ACRBTNA-H82E3200UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together