Biotinylated Human BAFF, Avitag,Fc Tag (active trimer) (MALS verified) (BAF-H82F3) is expressed from human 293 cells (HEK293). It co ntains AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 25 µG
Catalog # ACRBBAF-H82F3-25UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together