Order Entry
France
ContactUsLinkComponent
Human TNF-alpha, His Tag (active trimer) (MALS verified) (TNA-H5228) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession NP000585.2). 1 * 100 µG
Catalog # ACRBTNA-H5228100UG
Supplier:  ACROBIOSYSTEMS
Human TNF-alpha, His Tag (active trimer) (MALS verified) (TNA-H5228) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession NP000585.2). 1 * 100 µG
Catalog # ACRBTNA-H5228100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together