Order Entry
France
ContactUsLinkComponent
Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It conta ins AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 200 µG
Catalog # ACRBBAF-H82Q2200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It conta ins AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 200 µG
Catalog # ACRBBAF-H82Q2200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together