Order Entry
France
ContactUsLinkComponent
Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gl y 115 - Leu 260 (Accession P27548-1). 1 * 100 µG
Catalog # ACRBCDL-M5248100UG
Fournisseur:  ACROBIOSYSTEMS
Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gl y 115 - Leu 260 (Accession P27548-1). 1 * 100 µG
Catalog # ACRBCDL-M5248100UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together