Recombinat Protein,Human IgG2 Fc Protein, Tag Free,Human IgG Fc, Tag Free (IG2-H5206) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 326 (Accession P01859).This protein carries no tag. The protein has a calculated MW of 25.7 1 * 200 µG
Catalog # ACRBIG2-H5206200UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together