Order Entry
France
ContactUsLinkComponent
Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (CDL-H5266) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession P32970-1) trimer Design. 1 * 1 mg
Catalog # ACRBCDL-H5266-1MG
Supplier:  ACROBIOSYSTEMS
Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (CDL-H5266) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession P32970-1) trimer Design. 1 * 1 mg
Catalog # ACRBCDL-H5266-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together