Order Entry
France
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 389.1 MPA.S (25.00°C), RTM20 1 * 500 mL
Catalog # PARGRTM20
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 389.1 MPA.S (25.00°C), RTM20 1 * 500 mL
Catalog # PARGRTM20
Supplier:  PARAGON SCIENTIFIC
Frequently Bought Together