Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 75.19 MPA.S (25.00C), RTM13 1 * 500 mL
Catalog # PARGRTM13
Supplier:
PARAGON SCIENTIFIC
Expand image
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 75.19 MPA.S (25.00C), RTM13 1 * 500 mL
Catalog # PARGRTM13
Supplier:
PARAGON SCIENTIFIC
Frequently Bought Together
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals