Order Entry
France
ContactUsLinkComponent
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 48.59 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN26
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 48.59 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN26
Supplier:  PARAGON SCIENTIFIC