Order Entry
France
ContactUsLinkComponent
Spatula Set Contents: 2 spoon spatulas, 150 mm and 180 mm 3 micro-spoon spatulas, spoon 5 x 7 mm, L 100, 150, 185 mm 3 double spatulas, rigid, L 130, 150, 185 mm 1 micro-powder spatula, flexible, 150 mm 1 chemist's spoon, double-ended, 170 mm 1 spoon 1 * 1 SET
Catalog # ROTHKAT8.1
Supplier:  Roth Carl
Spatula Set Contents: 2 spoon spatulas, 150 mm and 180 mm 3 micro-spoon spatulas, spoon 5 x 7 mm, L 100, 150, 185 mm 3 double spatulas, rigid, L 130, 150, 185 mm 1 micro-powder spatula, flexible, 150 mm 1 chemist's spoon, double-ended, 170 mm 1 spoon 1 * 1 SET
Catalog # ROTHKAT8.1
Supplier:  Roth Carl
Frequently Bought Together