Order Entry
France
ContactUsLinkComponent
for sliding ring stirrer seal Ceramics / High Performance Graphite Shaft-Ø 10mm Seal Viton / integral spring ***Attention: - no liability for improper installation **** 1 * 1 items
Catalog # HWSA3792010
Supplier:  HWS LABORTECHNIK
for sliding ring stirrer seal Ceramics / High Performance Graphite Shaft-Ø 10mm Seal Viton / integral spring ***Attention: - no liability for improper installation **** 1 * 1 items
Catalog # HWSA3792010
Supplier:  HWS LABORTECHNIK
Frequently Bought Together