HYPERSIL PREPXL HS PHENYL 5UM 250X25MM PREP COLUMN 1 * 1 items
HYPE41505-259270XL
HYPERSIL PREPXL HS PHENYL 5UM 250X25MM PREP COLUMN 1 * 1 items
HYPE41505-259270XL
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection