Order Entry
France
ContactUsLinkComponent
CR-400 Chroma Meter Head including: CR-A43 White Calibration Plate CR-A72 Protective Cap 8 mm CR-A73 Wrist Strap AC-A305 AC-Adapter 1 * 1 items
Catalog # MINO1878-864
Supplier:  MINOLTA BUSINESS
CR-400 Chroma Meter Head including: CR-A43 White Calibration Plate CR-A72 Protective Cap 8 mm CR-A73 Wrist Strap AC-A305 AC-Adapter 1 * 1 items
Catalog # MINO1878-864
Supplier:  MINOLTA BUSINESS
Frequently Bought Together