SUCROSE TOTAL ORGANIC 0.5 MG/L C USP 1 * 125 mL
Catalog # REGEISTOC1185
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Laboratory trays
Supplier: KARTELL
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals