Order Entry
France
ContactUsLinkComponent
Segosoft is a certified software with a retraceable and unchangeable documentation of cycles. Further features: Networkable Password protected user accounts and Digital signature. The user has to buy the software and a yearly service card (SHPS30000 1 * 1 items
Catalog # SHPS300000600000
Fournisseur:  SHP Steriltechnik
Segosoft is a certified software with a retraceable and unchangeable documentation of cycles. Further features: Networkable Password protected user accounts and Digital signature. The user has to buy the software and a yearly service card (SHPS30000 1 * 1 items
Catalog # SHPS300000600000
Fournisseur:  SHP Steriltechnik
Frequently Bought Together