DUKE STANDARDS 2.0UM NIST TRACEABLE 1 * 15 mL
Catalog # MSPP4202A
Supplier: MARKET SOURCE PART PROCESS
DUKE STANDARDS 2.0UM NIST TRACEABLE 1 * 15 mL
Catalog # MSPP4202A
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Supplier: Merck
Diisodecyl phthalate, Sigma-Aldrich®
Supplier: Merck
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Butoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2,4-Trichlorobenzene ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific