Order Entry
France
ContactUsLinkComponent
L-lactate is produced from pyruvate by the enzyme Lacate Dehydrogenase. Lactate production occurs during anaerobic glycolysis or in proliferatively active cells. S 1 * 50 g
Catalog # GENORC-820
Supplier:  G-Biosciences
L-lactate is produced from pyruvate by the enzyme Lacate Dehydrogenase. Lactate production occurs during anaerobic glycolysis or in proliferatively active cells. S 1 * 50 g
Catalog # GENORC-820
Supplier:  G-Biosciences
Frequently Bought Together