Order Entry
France
ContactUsLinkComponent
Sodium Oxamate is a known glycolysis inhibitor. It inhibits the conversion of pyruvate to lactate via lactate dehydrogenase (LDH). S odium oxamate has been studied as an inhibitor of carbohydrate metabolism in various tumors, as cancer cells are ofte 1 * 25 g
Catalog # GENORC-816
Supplier:  G-Biosciences
Sodium Oxamate is a known glycolysis inhibitor. It inhibits the conversion of pyruvate to lactate via lactate dehydrogenase (LDH). S odium oxamate has been studied as an inhibitor of carbohydrate metabolism in various tumors, as cancer cells are ofte 1 * 25 g
Catalog # GENORC-816
Supplier:  G-Biosciences
Frequently Bought Together