NEEDLE HOLDER 73MM LUER-LOCK FOR 860 1 * 1 items
Catalog # METR6.2049.050
Supplier: METROHM
Frequently Bought Together
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
O-RINGS FOR NOSE CONE 1 * 24 items
Supplier: EPPENDORF
Accessories for Transferpette® electronic pipettes
Supplier: Brand
Accessories for Research® Plus pipettes
Supplier: EPPENDORF
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: Brand
Filter,membrane for research pipette 1 * 50 items
Supplier: EPPENDORF
Accessories for accu-jet® pro
Supplier: Brand
Accessories for Dispensette®
Supplier: Brand
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
FILTER FOR DRYING AGGREGAT 1 * 1 items
Supplier: MIELE
Accessories for PIPETBOY acu / pro
Supplier: Integra
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
Accessories for Rotavapor® R-3 / R-II / R-210 / R-215
Supplier: BUCHI