Order Entry
France
ContactUsLinkComponent
ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211) is expressed from human 293 cells (H EK293). It contains AA Val 77 - Leu 233 (Accession P01375-1). 1 * 100 µG
Catalog # ACRBTNA-H4211100UG
Supplier:  ACROBIOSYSTEMS
ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211) is expressed from human 293 cells (H EK293). It contains AA Val 77 - Leu 233 (Accession P01375-1). 1 * 100 µG
Catalog # ACRBTNA-H4211100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together