GREM2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human GREM2 in cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5174
Supplier: Abnova
Frequently Bought Together