Dll4 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat DLL4 in cell culture supernat es, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5196
Supplier: Abnova
Frequently Bought Together