Order Entry
France
Orders LinkContactUsLinkComponent
Sterilisation flat pouches, Stericlin®
Sterilisation flat pouches, Stericlin®
Sterilisation pouches, with indicator, 200×400 mm
Catalog # 129-1299
Sterilisation flat pouches, Stericlin®
Catalog # 129-1299
Sterilisation flat pouches, Stericlin®
Sterilisation pouches, with indicator, 200×400 mm
Price
627,00 €
Log in for availability
1 * 500 Pce
Product Details & Documents
Stericlin® sterilisation pouches, Tyvek® 1073B with see-through film and indicators for VH₂O₂ (plasma) and EO.
  • Nonwoven polyethylene
Barrier properties against germs and moisture, extreme high puncture resistance and fiber-free peel without delamination draw this sterile barrier system. Suitable for steam sterilisation, but absolutely compatible with the VH₂O₂ (plasma) and radiation sterilisation.
Tyvek® and the composite film used are ideal for VH₂O₂ (plasma), EO, FORM and irradiation sterilisation. Two indicators for the VH₂O₂ (plasma) and EO sterilisation are printed outside the filling area.
The stericlin® sterile barrier system is first choice in gas sterilisation, as Tyvek® and the composite film hardly absorb the toxic gases and enable residual gases to be far below specified limits.
Certifications: DIN EN ISO 11607 and DIN 868-5.
Specifications
  • Description:
    Sterilisation pouches, with indicator
  • L×W:
    200×400 mm
  • Seq:
    04