Ammonia solution 25% 100 ml/l solution
10 L
Fournisseur: PROLABO CMP
Ammonia solution 25% 100 ml/l solution
Référence PRLS85163.410
Fournisseur: PROLABO CMP
Numéro CAS: 1336-21-6
Jump to:
Product Details & Documents
Concentrated solution to dilute to 100 mL/L.
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Density:0.92 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- ADR:8,III
Specifications
Specifications
- Pk:10 L



