Order Entry
France
Orders LinkContactUsLinkComponent
Mouse IgM purification kit

Mouse IgM purification kit

LigaTrap® Mouse IgM purification column, 1×1 ml, 1 KIT
Catalog # LIGALT-145-1X1ML
Marketsource
Mouse IgM purification kit
Catalog # LIGALT-145-1X1ML
Marketsource
CAS Number:  
Mouse IgM purification kit

LigaTrap® Mouse IgM purification column, 1×1 ml, 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents

LigaTrap® Mouse IgM Purification Kit with LT technology utilises novel, patented ligands, designed for IgM antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of mouse IgM in just ten easy steps.

  • Designed to purify mouse IgM
  • For in vitro laboratory use
  • Binding capacity for mouse IgM is >10 mg/ml resin
  • Storage: 2 to 8 °C

LigaTrap® mouse IgM purification resin is engineered to purify high quality mouse IgM antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.

Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.

Delivery information: Each kit contains ten microspin columns prefilled with 0,1 ml of LigaTrap® mouse IgM purification resin, buffers and collection tubes.

Caution: Not for diagnostic or therapeutic use. Not for human or animal consumption.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    LigaTrap® Mouse IgM purification column, 1×1 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.