Order Entry
France
ContactUsLinkComponent
Apparatus for the determination of etheric oils Made of borosilicate glass 3.3. acc. to DAB. Complete distillation head with ground glass cone 29/32, Allihn cooler, vent plug NS 7/16, 1 ml measuring tube (graduated 0.01 ml) with 3 ml ball, refill/ove 1 * 1 SET
Catalog # ROTHXX23.1
Supplier:  Roth Carl
Apparatus for the determination of etheric oils Made of borosilicate glass 3.3. acc. to DAB. Complete distillation head with ground glass cone 29/32, Allihn cooler, vent plug NS 7/16, 1 ml measuring tube (graduated 0.01 ml) with 3 ml ball, refill/ove 1 * 1 SET
Catalog # ROTHXX23.1
Supplier:  Roth Carl
Frequently Bought Together