CTSL (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human Cathepsin L in cell cultu re supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5059
Supplier: Abnova
Frequently Bought Together