Order Entry
France
ContactUsLinkComponent
Rotilabo-syringe filters, sterile, PTFE, Ø 25 mm 0,20µm 100pcs/pck 1 * 100 items
Catalog # ROTHKC76.1
Supplier:  Roth Carl
Rotilabo-syringe filters, sterile, PTFE, Ø 25 mm 0,20µm 100pcs/pck 1 * 100 items
Catalog # ROTHKC76.1
Supplier:  Roth Carl
Frequently Bought Together