MIKRO-SCHRAUBRÖHRE 1.5ML PROTEIN LOW BINDING DECKEL VERSCHRAUBT PCR-PT TYP
Catalog # SARS72.703.600
Supplier: SARSTEDT
MIKRO-SCHRAUBRÖHRE 1.5ML PROTEIN LOW BINDING DECKEL VERSCHRAUBT PCR-PT TYP
Catalog # SARS72.703.600
Supplier: SARSTEDT
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Microtome blades, disposable, FEATHER®
Supplier: PFM Medical GmbH
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Microcentrifuge Tubes with Flat Screw Caps
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
TUBE MICRO SCREW 0.5ML PP LID 1 * 1.000 items
Supplier: SARSTEDT